Membrane filter funnels, CN, Microsart® @filter
Supplier: Sartorius
16A08 16D01-10-07-TG 16D01-10-H6-BL 16D01-10-07-BL 16D01-25-H6-TF 16D02-10-06-TG 16D03-25-H6-TF 16D03-25-05-BK 16D03-25-H6-BK 16D02-10-06-BL 16D02-25-06-BK 16D02-25-06-TF 16D03-10-H6-BL 16D03-10-H6-TG 16D01-10-H6-TG 16D01-25-07-BK 16D01-25-07-TF 16D01-25-H6-BK 16D01-10-06-TG 16D01-10-06-BL 16D01-25-06-BK 16D01-10-H6ACG 16D01-25-H6ACF 16D01-25-07ACF 16D03-25-H6ACF 16D03-10-H6ACG 16D01-10-07ACG
516-0420EA 1380 EUR
516-0420 516-0393 516-0399 516-0398 516-0404 516-0396 516-0405 516-0412 516-0410 516-0401 516-0411 516-0406 516-0400 516-0395 516-0394 516-0408 516-0403 516-0409 SART16D01-10-06-TG SART16D01-10-06-BL SART16D01-25-06-BK 516-1163 516-1165 516-1164 516-1167 516-1166 SART16D01-10-07ACG
Membrane filter funnels, CN, Microsart® @filter
Funnels Membrane Filter Funnels
These filter units with PP funnel and base and cellulose nitrate membrane are developed for the detection and enumeration of microorganisms in pharmaceuticals, cosmetics, food, beverages, water and other liquids. All components of the filtration system must comply with international guidelines, such as USP, EP or ISO standards.
  • No liquid remains after filtration, no need for rinsing
  • Click-Fit closure, no risk of leakage
  • Adaptable on Combisart®
  • Transparent funnel material with marked graduations
Microsart® Base 47 mm is the perfect addition to existingMore Product Information