Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Supplier: DWK Life Sciences
SP179950-4532
SCERSP179950-4532EA 384 EUR
SCERSP179950-4532
Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Filters Membrane Filter Units
This microfiltration adapter assembly is designed to allow connection of a 47 or 90 mm filtration support base with a straight drip tip to a bottle or reservoir with a GL 45 thread.
Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly.
Certifikace: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements