Petrochemical filter stick apparatus with hooks and springs
Supplier: DWK Life Sciences
SP513880-0000
SCERSP513880-0000EA 311 EUR
SCERSP513880-0000
Petrochemical filter stick apparatus with hooks and springs
Petroleum Testing Apparatus
The Filter Stick Apparatus is used in the determination of oil content and solvent extractables in petroleum waxes.
  • Supplied with one pair of springs
  • Designed from ASTM D721 specifications
  • Manufactured from borosilicate 3,3 glass
Assembly consists of a sintered glass filter stick with air pressure inlet tube and delivery nozzle and a cooling tube with NS 24/40 joint.
Certifikace: The borosilicate glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.