Tissue grinders, Pellet Pestle®, KIMBLE®
Supplier: DWK Life Sciences
SP749515-0000 SP749516-0500
SCERSP749515-0000EA 316 EUR
SCERSP749515-0000 SCERSP749516-0500
Tissue grinders, Pellet Pestle®, KIMBLE®
Homogenisers Tissue Grinders
Pellet pestles with a stainless steel shaft/CTFE tips.
  • Reusable pellet pestles for microcentrifuge and centrifuge tubes