Tissue grinders, Tenbroeck type, KIMBLE®
Supplier: DWK Life Sciences
SP885000-0040 SP885000-0015 SP885000-0002 SP885000-0007
SCERSP885000-0040EA 489 EUR
SCERSP885000-0040 SCERSP885000-0015 SCERSP885000-0002 SCERSP885000-0007
Tissue grinders, Tenbroeck type, KIMBLE®
Homogenisers Tissue Grinders
Borosilicate glass with ground glass pestle.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • For hand or motor operation
  • Replacement parts are completely interchangable
  • Clearance between pestles and tubes is 0,10 to 0,15 mm
The pestle is tooled for a single hole rubber stopper. By inserting one end of a short metal rod in the stopper, these grinders can be motor driven at slow speeds by inserting the other end of the metal rod into the motor chuck. Ideal forMore Product Information