Ice Flakers, Self Contained, AF
Supplier: SCOTSMAN
AF80AS AF156 AF124 AF127
SCOMAF80ASEA 3890 EUR
SCOMAF80AS 474-0112 474-0111 SCOMAF127
Ice Flakers, Self Contained, AF
Ice Makers
The AF self contained ice flakers are air-cooled and manufactured from 304 stainless steel.
  • Integral ice flake storage bin
  • Constructed of high grade stainless steel
  • Ideal for medical and laboratory use
All the models are automatic and produce small irregular flakes of ice with an even melting rate. The flakes are easily moulded to fit into any shape. The ice flakers require a 19 mm 'washing machine' type tapped mains water supply, a correctly rated power supply and a 38 mm vented drain (ideally no higher than the base of the machine), within one metreMore Product Information