Staining jars, DURAN®
Supplier: DWK Life Sciences
233160003
SCOT233160003EA 197 EUR
SCOT233160003
Staining jars, DURAN®
Jars Staining Jars
Soda-lime glass.
  • For 10 slides
These staining jars are suitable for standard microscope slides (76×26 mm).
Upozornení: Note: Do not clean staining dishes and jars at temperatures above 60°C (glass corrosion is possible).