Measuring Pipettes, Class AS
Supplier: DWK Life Sciences
233470607 233471106 233471603 233472308 233472908 233473201 233473407 233473604 233490609 233491108 233491605 233492301 233492901 233493203 233493409 233493606
SCOT233470607EA 94.6 EUR
SCOT233470607 SCOT233471106 SCOT233471603 SCOT233472308 SCOT233472908 SCOT233473201 SCOT233473407 SCOT233473604 SCOT233490609 SCOT233491108 SCOT233491605 SCOT233492301 SCOT233492901 SCOT233493203 SCOT233493409 SCOT233493606
Measuring Pipettes, Class AS
Pipettes Measuring (Mohr) Pipettes
Pipettes are used for precise measurement and decanting of liquids. Graduated pipettes allow different amounts of liquid to be absorbed and dispensed in the same or in different sizes. The tolerances for the volume content of calibratable pipettes correspond to the error limits of the German weights and measures regulations as well as ISO and DIN.
  • Manufactured from soda-lime glass
  • Rapid discharge reduces significantly the waiting time compared to Class A
  • Calibration is based on the poured out volume ('Ex') at a +20 °C reference temperature
  • With circular