SafetyCaps, V2.0, GL 45, with shut-off valve
Supplier: S.C.A.T.
307919 307920 307519 307219 307319 307419
SCTV307919EA 151 EUR
SCTV307919 SCTV307920 SCTV307519 SCTV307219 SCTV307319 SCTV307419
SafetyCaps, V2.0, GL 45, with shut-off valve
Closures Safety Caps and Safety Waste Caps
These SafetyCaps are designed to safely extract solvents when working with any HPLC/UHPLC system.
  • Easy change of supply containers during the analysis process
  • No contamination of the solvent
  • No risk of air pockets in capillaries
After the supply container is changed, the current analysis can be continued without interruption.
During repairs on HPLC pumps, the shut-off on the SafetyCaps can be closed so the capillaries and solvent filter remain in the solvent. Thus, the pump can be quickly and easily flushed when returning to operation after repairs.