Safety Waste Cap "LISA", V3.0
Supplier: S.C.A.T.
350045 350050 350051 350053 350055 350060 350063 350065 350070 350083 350090 350095 450045 450060 350100 450100 350120 450120 450055 350122 450050 450070 450090 450121 450122
SCTV350045EA 164 EUR
SCTV350045 SCTV350050 SCTV350051 SCTV350053 SCTV350055 SCTV350060 SCTV350063 SCTV350065 SCTV350070 SCTV350083 SCTV350090 SCTV350095 SCTV450045 SCTV450060 SCTV350100 SCTV450100 SCTV350120 SCTV450120 SCTV450055 SCTV350122 SCTV450050 SCTV450070 SCTV450090 SCTV450121 SCTV450122
Safety Waste Cap "LISA", V3.0
Closures Safety Caps and Safety Waste Caps
The Safety Waste Caps LISA’s V3.0 high quality innovative docking system of the, is the latest generation of a modern and universally expandable modular system for the safe disposal of liquid HPLC / UHPLC waste. The disposal system offers a high number of individual threads, connections and expansion options.
  • Satellite blind plug - for extending and attaching various components such as a funnel
  • Tube connector - for connecting larger tubes with 5 to 11,5 mm ID
  • Satellite made of PTFE - for optimal chemical resistance
  • 360° freely rotatable screw cap - for