Disinfectant, terralin® Liquid
Supplier: SCHUELKE MAYR
102001
SCUE102001EA 249 EUR
SCUE102001
Disinfectant, terralin® Liquid
Cleaning Agents Disinfectants
terralin® can be used for rapid disinfection of medical devices [CE0297] in all hospital areas with a high infection risk, e.g. treatment units and adjoining surfaces, examination tables, operating tables and adjoining surfaces.
  • Alcohol based rapid disinfectant, containing no aldehydes
  • Ready to use
  • Rapid and effective in many applications
  • Dries rapidly, leaving no streaks
Order Now
SPECIFICATIONS
Model Terralin® liquid
Environmentally Preferable
Bactericidal activity MRSA, Tuberculosis
Virucidal activity Hepatitis B viruses, HIV, HCV, adenoviruses, rotaviruses, papovaviruses, polioviruses
Ingredients Ethanol 25%, Propan-1-ol 35%
Chemical and physical specifications clear, colourless liquid, SG=0,895; pH value= 6