Silicone inserts for pipette containers
Supplier: schuett-biotec GmbH
3523213
SCUT3523213EA 9.6 EUR
SCUT3523213
Silicone inserts for pipette containers
Inserts
Silicone rubber for insertion in VARICON RS (lower part) and SPECICON (lower and upper part) pipette containers and it is designed to prevent damage to pipettes caused by sudden shock.
Thickness: 2 mm
Ø: 58 mm