Buckets Square
Supplier: SEMADENI
1358
SEMA1358EA 106 EUR
SEMA1358
Buckets Square
Buckets
Buckets made of LDPE, translucent, for industrial and laboratory applications.
  • Heavy
  • Sturdy
  • With internal scale and metal carrying handle