High pressure syringes
Supplier: Trajan Scientific and Medical
008170 008171
SGEA008170EA 290 EUR
SGEA008170 SGEA008171
High pressure syringes
Syringes Syringes for Chromatography
These high pressure syringes are designed for high pressure gas sampling.
  • Rated to 34 bar
  • Fitted with a gas tight on/off valve and high pressure 70 mm, 23 gauge needle