CRS ONE O-Rings for Inlet Liner Seals
Supplier: Trajan Scientific and Medical
0726782 0726783 0726786
SGEA0726782EA 23 EUR
SGEA0726782 SGEA0726783 SGEA0726786
CRS ONE O-Rings for Inlet Liner Seals
Seals Sealing Rings
The o-ring prevents gas leaks and maintains a uniform flow path through the inlet, where even small leaks can cause sample loss - affecting accuracy and reproducibility of results.
  • Consistently forms a reliable seal, preventing inlet leaks and sample loss
  • Pre-cleaned and conditioned to minimize potential active sites and system contamination
  • Resistant to high temperatures and chemical exposure, ensuring optimal performance, durability and longevity
  • Compatible with multiple instruments
Applications: GC and GC-MS analysis. Ideal for trace level analysisMore Product Information