Trisodium phosphate dodecahydrate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate

M-Clarity™ quality level = MQ200

04277-1KG 04277-2.5KG 04277-25KG-H 04277-25KG
SIAL04277-1KGEA 40.8 EUR
SIAL04277-1KG SIAL04277-2.5KG SIAL04277-25KG-H SIAL04277-25KG
Trisodium phosphate dodecahydrate, Sigma-Aldrich®
tri-Sodium phosphate dodecahydrate
Formula: Na₃PO₄·12H₂O
MW: 380.12 g/mol
Melting Pt: ∼75 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR