2,5-Xylidine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2-Amino-p-xylene, 2,5-Dimethylaniline

M-Clarity™ quality level = MQ100

102253-100ML
SIAL102253-100MLEA 53.7 EUR
SIAL102253-100ML
2,5-Xylidine, Sigma-Aldrich®
2,5-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 15 °C
Density: 0.979 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007743
CAS Number: 95-78-3
EINECS: 202-451-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR