2-Acetylnaphthalene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2´-Acetonaphthone, 1-(Naphthalen-2-yl)ethanone, Methyl β-naphthyl ketone, 2-Acetonaphthone, 2'-Acetonaphthone

M-Clarity™ quality level = MQ200

134775-100G 134775-5G
SIAL134775-100GEA 245 EUR
SIAL134775-100G SIAL134775-5G
2-Acetylnaphthalene, Sigma-Aldrich®
2-Acetylnaphthalene
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300
Melting Pt: 52…54 °C
Density: 1.1171 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR