4-Methyl-3-nitroanisole, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Methoxy-4-methyl-3-nitrobenzene, 4-Methoxy-2-nitrotoluene, Methyl 3-nitro-p-tolyl ether

M-Clarity™ quality level = MQ100

139823-5G
SIAL139823-5GEA 64 EUR
SIAL139823-5G
4-Methyl-3-nitroanisole, Sigma-Aldrich®
4-Methyl-3-nitroanisole
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007173
CAS Number: 17484-36-5
EINECS: 241-500-0
UN: 2446
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR