Pentamethylbenzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,2,3,4,5-Pentamethylbenzene

153613-100G
SIAL153613-100GEA 293 EUR
SIAL153613-100G
Pentamethylbenzene, Sigma-Aldrich®
Pentamethylbenzene
Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 51…52 °C
Density: 0.917 g/cm³ (20 °C)
MDL Number: MFCD00008522
CAS Number: 700-12-9
EINECS: 211-837-8
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR