N-Boc-ethylenediamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-(tert-Butoxycarbonyl)-1,2-diaminoethane, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl (2-aminoethyl)carbamate, t-boc-N-EDA, tert-Butyl N-(2-Aminoethyl)carbamate, N-BOC-1,2-diaminoethane

15369-1G 15369-25G 15369-5G
SIAL15369-1GEA 132 EUR
SIAL15369-1G SIAL15369-25G SIAL15369-5G
N-Boc-ethylenediamine, Sigma-Aldrich®
N-Boc-ethylenediamine
Formula: C₇H₁₆N₂O₂
MW: 160.22 g/mol
MDL Number: MFCD00191871
CAS Number: 57260-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR