3-Methyl-2-buten-1-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methyl-2-buten-4-ol

M-Clarity™ quality level = MQ100

162353-100ML 162353-500ML 162353-5ML
SIAL162353-100MLEA 63.2 EUR
SIAL162353-100ML SIAL162353-500ML SIAL162353-5ML
3-Methyl-2-buten-1-ol, Sigma-Aldrich®
3-Methyl-2-buten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 140
Melting Pt: –86 °C
Density: 0.848 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002916
CAS Number: 556-82-1
EINECS: 209-141-4
UN: 1987
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR