2-Chloro-2-methylpropane, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: tert-Butyl chloride

MQ200

19780-1L 19780-250ML
SIAL19780-1LEA 80.4 EUR
SIAL19780-1L SIAL19780-250ML
2-Chloro-2-methylpropane, Sigma-Aldrich®
2-Chloro-2-methylpropane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 51
Melting Pt: –28 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000816
CAS Number: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3 II
Merck Index: 13,01561

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR