Tin(IV) chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Stannic chloride, Tin tetrachloride

M-Clarity™ quality level = MQ200

208930-250G 208930-2KG
SIAL208930-250GEA 68.6 EUR
SIAL208930-250G SIAL208930-2KG
Tin(IV) chloride, Sigma-Aldrich®
Tin (IV) chloride
Formula: Cl₄Sn
MW: 260.52 g/mol
Boiling Pt: 114
Melting Pt: –33 °C
Density: 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8 II
Merck Index: 12,08929

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR