5-Hydroxy-1-tetralone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one

M-Clarity™ quality level = MQ100

219975-5G
SIAL219975-5GEA 48.1 EUR
SIAL219975-5G
5-Hydroxy-1-tetralone, Sigma-Aldrich®
5-Hydroxy-1-tetralone
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00001693
CAS Number: 28315-93-7
EINECS: 248-958-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR