2-Nitrocyclohexanone, Sigma-Aldrich®

Supplier: Merck

224480-5G
SIAL224480-5GEA 240 EUR
SIAL224480-5G
2-Nitrocyclohexanone, Sigma-Aldrich®
2-Nitrocyclohexanone
Formula: C₆H₉NO₃
CAS Number: 4883-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR