3,4,5-Trimethoxytoluene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,2,3-Trimethoxy-5-methylbenzene

M-Clarity™ quality level = MQ100

227714-10G
SIAL227714-10GEA 82.6 EUR
SIAL227714-10G
3,4,5-Trimethoxytoluene, Sigma-Aldrich®
3,4,5-Trimethoxytoluene
Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 35…37 °C
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00008397
CAS Number: 6443-69-2
EINECS: 229-239-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR