2-Chloroaniline, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

23310-250ML
SIAL23310-250MLEA 55 EUR
SIAL23310-250ML
2-Chloroaniline, Sigma-Aldrich®
2-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.213 g/cm³ (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00007656
CAS Number: 95-51-2
EINECS: 202-426-4
UN: 2019
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR