Zinc, Sigma-Aldrich®

Supplier: Merck


MQ200

243469-100G 243469-500G 243469-2KG
SIAL243469-100GEA 41.5 EUR
SIAL243469-100G SIAL243469-500G SIAL243469-2KG
Zinc, Sigma-Aldrich®
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR