8-Methylquinoline, Sigma-Aldrich®

Supplier: Merck
Warning


MQ100

261769-5G
SIAL261769-5GEA 76.7 EUR
SIAL261769-5G
8-Methylquinoline, Sigma-Aldrich®
8-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 248
Melting Pt: –80 °C
Density: 1.0852 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006810
CAS Number: 611-32-5
EINECS: 210-264-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR