Cobalt, Sigma-Aldrich®

Supplier: Merck
Danger


MQ100

266701-1.4G
SIAL266701-1.4GEA 325 EUR
SIAL266701-1.4G
Cobalt, Sigma-Aldrich®
Cobalt
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR