Pyridoxal hydrochloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl

M-Clarity™ quality level = MQ200

271748-1G 271748-25G 271748-5G
SIAL271748-1GEA 43.9 EUR
SIAL271748-1G SIAL271748-25G SIAL271748-5G
Pyridoxal hydrochloride, Sigma-Aldrich®
Pyridoxal hydrochloride
Formula: C₈H₁₀ClNO₃
MW: 203.62 g/mol
MDL Number: MFCD00012809
CAS Number: 65-22-5
EINECS: 200-602-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR