Indium, powder, Sigma-Aldrich®

Supplier: Merck


MQ100

277959-10G 277959-50G
SIAL277959-10GEA 149 EUR
SIAL277959-10G SIAL277959-50G
Indium, powder, Sigma-Aldrich®
Indium
Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.31 g/cm³ (20 °C)
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4.1 III
Merck Index: 13,04971

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR