Crotonaldehyde stabilised, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Butenal, mixture of cis and trans, Crotonaldehyde, mixture of cis and trans

27980-250ML
SIAL27980-250MLEA 149 EUR
SIAL27980-250ML
Crotonaldehyde stabilised, Sigma-Aldrich®
Crotonaldehyde
Formula: C₄H₆O
MW: 70.09 g/mol
MDL Number: MFCD00007003
CAS Number: 4170-30-3
EINECS: 224-030-0
UN: 1143
ADR: 6.1 I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR