Dipropyleneglycol monomethyl ether (mixture of isomers), Sigma-Aldrich®

Supplier: Merck

Synonyms: Dipropylene glycol monomethyl ether, 1-((1-Methoxypropan-2-yl)oxy)propan-2-ol

MQ200

283282-3L 283282-18L 283282-1L
SIAL283282-3LEA 136 EUR
SIAL283282-3L SIAL283282-18L SIAL283282-1L
Dipropyleneglycol monomethyl ether (mixture of isomers), Sigma-Aldrich®
Dipropyleneglycol monomethyl ether (mixture of isomers)
Formula: C₇H₁₆O₃
MW: 148.2001 g/mol
MDL Number: MFCD00059604
CAS Number: 34590-94-8
EINECS: 252-104-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR