3,5-Diiodosalicylaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3,5-Diiodo-2-hydroxybenzaldehyde

M-Clarity™ quality level = MQ200

283444-5G
SIAL283444-5GEA 59.4 EUR
SIAL283444-5G
3,5-Diiodosalicylaldehyde, Sigma-Aldrich®
3,5-Diiodosalicylaldehyde
Formula: C₇H₄I₂O₂
MW: 373.92 g/mol
MDL Number: MFCD00003321
CAS Number: 2631-77-8
EINECS: 220-117-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR