3-Methoxyphenylacetone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-(3-Methoxyphenyl)propan-2-one

M-Clarity™ quality level = MQ100

288810-1G
SIAL288810-1GEA 61.6 EUR
SIAL288810-1G
3-Methoxyphenylacetone, Sigma-Aldrich®
3-Methoxyphenylacetone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00008771
CAS Number: 3027-13-2
EINECS: 221-191-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR