Styrene-[D₈] Stabilised, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Phenylethene-D8, Styrene-[²H₈], Vinylbenzene-D8, Phenylethylene-D8

Stabilization: 0.5 % 4-tert-Butylbrenzcatechin
MQ200

338222-1G 338222-5G
SIAL338222-1GEA 203 EUR
SIAL338222-1G SIAL338222-5G
Styrene-[D₈] Stabilised, Sigma-Aldrich®
Styrene-[D8]
Formula: C₆D₅CDCD₂
MW: 112.09 g/mol
Boiling Pt: 145
Melting Pt: –31 °C
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00044231
CAS Number: 19361-62-7
EINECS: 242-995-6
UN: 2055
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR