3,4,5-Trimethoxybenzylamine, Sigma-Aldrich®

Supplier: Merck


MQ100

338931-25G 338931-5G
SIAL338931-25GEA 201 EUR
SIAL338931-25G SIAL338931-5G
3,4,5-Trimethoxybenzylamine, Sigma-Aldrich®
3,4,5-Trimethoxybenzylamine
Formula: C₁₀H₁₅NO₃
MW: 197.23
Boiling Pt: 106 °C (0.1 mmHg)
Density: 1.155 g/cm³ (20 °C)
MDL Number: MFCD00017150
CAS Number: 18638-99-8
EINECS: 242-468-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR