N-Methylphthalimide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methylisoindoline-1,3-dione

M-Clarity™ quality level = MQ100

407992-5G
SIAL407992-5GEA 45.9 EUR
SIAL407992-5G
N-Methylphthalimide, Sigma-Aldrich®
N-Methylphthalimide
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 134 °C
MDL Number: MFCD00023063
CAS Number: 550-44-7
EINECS: 208-982-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR