Methanol-[D₄], Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

MQ200

422878-10ML 422878-50ML
SIAL422878-10MLEA 171 EUR
SIAL422878-10ML SIAL422878-50ML
Methanol-[D₄], Sigma-Aldrich®
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.3
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR