4-Bromo-3-nitrotoluene, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Bromo-4-methyl-2-nitrobenzene

M-Clarity™ quality level = MQ100

424870-25ML
SIAL424870-25MLEA 256 EUR
SIAL424870-25ML
4-Bromo-3-nitrotoluene, Sigma-Aldrich®
4-Bromo-3-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 84…100 °C (0.25 mmHg)
Melting Pt: 35 °C
Density: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00024180
CAS Number: 5326-34-1
EINECS: 226-203-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR