4-(Chloromethyl)styrene 90% (by GC) inhibited, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride

Stabilization: 500 ppm Inhibited with tert-butylcatechol
M-Clarity™ quality level = MQ200

436887-100ML 436887-25ML
SIAL436887-100MLEA 75.1 EUR
SIAL436887-100ML SIAL436887-25ML
4-(Chloromethyl)styrene 90% (by GC) inhibited, Sigma-Aldrich®
4-(Chloromethyl)styrene

Component of ion exchange resins, photo-resist polymers, cross-linkable fibers, coupling agents and electroconducting polymers.


Starter for various copolymer preparations. Dual functional monomer. Readily derivatised by chloride displacement.

Formula: C₉H₉Cl
MW: 152.62 g/mol
Boiling Pt: 229 °C (lit.)
Density: 1.083 g/lL (25 °C) (lit.)
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051362
CAS Number: 1592-20-7
EINECS: 216-471-2
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Infrared Spectrum Conforms to Structure
Purity (GC) ≥89.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR