(3-Bromopropyl)trichlorosilane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

437808-5ML
SIAL437808-5MLEA 97.6 EUR
SIAL437808-5ML
(3-Bromopropyl)trichlorosilane, Sigma-Aldrich®
(3-Bromopropyl)trichlorosilane
Formula: BrH₂CCH₂CH₂SiCl₃
MW: 256.43 g/mol
Boiling Pt: 50 °C (0.001 mmHg)
Density: 1.685 g/cm³ (20 °C)
MDL Number: MFCD00039291
CAS Number: 13883-39-1
EINECS: 237-650-1
UN: 2987
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR