5-Bromo-2-fluorobenzonitrile, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

465259-5G
SIAL465259-5GEA 27.4 EUR
SIAL465259-5G
5-Bromo-2-fluorobenzonitrile, Sigma-Aldrich®
5-Bromo-2-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 255
Melting Pt: 75…80 °C
MDL Number: MFCD00143424
CAS Number: 179897-89-3
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR