2,6-Dimethylphenylboronic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,6-Dimethylbenzeneboronic acid, (2,6-Dimethylphenyl)boronic acid, Dihydroxy(2,6-dimethylphenyl)borane, (2,6-Dimethylphenyl)metaboric acid

MQ100

480061-25G 480061-5G
SIAL480061-25GEA 151 EUR
SIAL480061-25G SIAL480061-5G
2,6-Dimethylphenylboronic acid, Sigma-Aldrich®
2,6-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD01009693
CAS Number: 100379-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR