Di-n-octyl phthalate-[ring-D₄] (98% D), Sigma-Aldrich®

Supplier: Merck

Synonyms: Di-n-octyl phthalate-[ring-²H₄]

M-Clarity™ quality level = MQ200

488887-100MG
SIAL488887-100MGEA 552 EUR
SIAL488887-100MG
Di-n-octyl phthalate-[ring-D₄] (98% D), Sigma-Aldrich®
Di-n-octyl phthalate-[ring-D4]
Formula: C₂₄H₃₄D₄O₄
MDL Number: MFCD00144150
CAS Number: 93952-13-7
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR