D-(+)-Glucose, Sigma-Aldrich®

Supplier: Merck

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

49139-1KG 49139-5KG 49139-250G
SIAL49139-1KGEA 121 EUR
SIAL49139-1KG SIAL49139-5KG SIAL49139-250G
D-(+)-Glucose, Sigma-Aldrich®
D(+)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR