3,4-Dimethylbenzaldehyde, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

493856-5G
SIAL493856-5GEA 103 EUR
SIAL493856-5G
3,4-Dimethylbenzaldehyde, Sigma-Aldrich®
3,4-Dimethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 68 °C (0.8 mmHg)
Density: 1.012 g/cm³ (20 °C)
MDL Number: MFCD00016612
CAS Number: 5973-71-7
EINECS: 227-770-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR