2-Hydroxyphenylboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

M-Clarity™ quality level = MQ100

522554-5G
SIAL522554-5GEA 211 EUR
SIAL522554-5G
2-Hydroxyphenylboronic acid pinacol ester, Sigma-Aldrich®
2-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
MDL Number: MFCD02093754
CAS Number: 269409-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR